Please login to your online account to display your discounted pricing

2-Fluoro-5-nitrobenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-nitrobenzoic acid
CAS 7304-32-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4FNO4
Linear Formula NO2C6H3(F)COOH
Formula Weight 185.11
MDL Number MFCD00134238
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398170010

  SDS
thermo fisher scientific
AC398170010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398170050

  SDS
thermo fisher scientific
AC398170050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 185.11
Physical Form Crystalline Powder
Quantity 1g
Color White to pale yellow
Melting Point 138° to 142°C
CAS Max % 100.0
Molecular Formula C7H4FNO4
Linear Formula NO2C6H3(F)COOH
Chemical Name or Material 2-Fluoro-5-nitrobenzoic acid
CAS 7304-32-7
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00134238
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.