missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-nitrobenzeneboronic acid, 97%

$67.42 - $276.32

Chemical Identifiers

CAS 819849-20-2
Molecular Formula C6H5BFNO4
Molecular Weight (g/mol) 184.917
MDL Number MFCD03092929
InChI Key SYAFEPQKPQIRAL-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitro phenyl boronic acid, 2-fluoro-5-nitro-phenyl boronic acid, 2-fluoro-5-nitrobenzeneboronic acid, 2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 5-nitro-2-fluorophenylboronic acid, acmc-209pmq, b-2-fluoro-5-nitrophenyl-boronic acid, boronic acid, 2-fluoro-5-nitrophenyl, ksc649e7n
PubChem CID 2763280
IUPAC Name (2-fluoro-5-nitrophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)[N+](=O)[O-])F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6410003 View Documents Alfa Aesar™
H6410003
1g Each for $67.42
 
AAH6410006 View Documents Alfa Aesar™
H6410006
5g Each for $276.32
 
Description

Description

Applications
2-Fluoro-5-nitrobenzeneboronic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

819849-20-2
184.917
SYAFEPQKPQIRAL-UHFFFAOYSA-N
2763280
B(C1=C(C=CC(=C1)[N+](=O)[O-])F)(O)O
C6H5BFNO4
MFCD03092929
2-fluoro-5-nitro phenyl boronic acid, 2-fluoro-5-nitro-phenyl boronic acid, 2-fluoro-5-nitrobenzeneboronic acid, 2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 5-nitro-2-fluorophenylboronic acid, acmc-209pmq, b-2-fluoro-5-nitrophenyl-boronic acid, boronic acid, 2-fluoro-5-nitrophenyl, ksc649e7n
(2-fluoro-5-nitrophenyl)boronic acid

Specifications

White
819849-20-2
MFCD03092929
Slightly soluble in water.
B(C1=C(C=CC(=C1)[N+](=O)[O-])F)(O)O
184.917
184.92
97%
1g
C6H5BFNO4
2-fluoro-5-nitro phenyl boronic acid, 2-fluoro-5-nitro-phenyl boronic acid, 2-fluoro-5-nitrobenzeneboronic acid, 2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 5-nitro-2-fluorophenylboronic acid, acmc-209pmq, b-2-fluoro-5-nitrophenyl-boronic acid, boronic acid, 2-fluoro-5-nitrophenyl, ksc649e7n
SYAFEPQKPQIRAL-UHFFFAOYSA-N
(2-fluoro-5-nitrophenyl)boronic acid
2763280
Powder
2-Fluoro-5-nitrobenzeneboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures