missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-nitrobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H6410006

 View more versions of this product

Catalog No. AAH6410006



Description

Description

Applications
2-Fluoro-5-nitrobenzeneboronic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Fluoro-5-nitrobenzeneboronic acid
5g
C6H5BFNO4
2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 2-fluoro-5-nitrobenzeneboronic acid, 5-nitro-2-fluorophenylboronic acid, boronic acid, 2-fluoro-5-nitrophenyl, 2-fluoro-5-nitro-phenyl boronic acid, acmc-209pmq, ksc649e7n, 2-fluoro-5-nitro phenyl boronic acid, b-2-fluoro-5-nitrophenyl-boronic acid
B(C1=C(C=CC(=C1)[N+](=O)[O-])F)(O)O
184.917
184.92
97%
White
819849-20-2
MFCD03092929
SYAFEPQKPQIRAL-UHFFFAOYSA-N
(2-fluoro-5-nitrophenyl)boronic acid
2763280
Powder

Chemical Identifiers

819849-20-2
184.917
SYAFEPQKPQIRAL-UHFFFAOYSA-N
2763280
B(C1=C(C=CC(=C1)[N+](=O)[O-])F)(O)O
C6H5BFNO4
MFCD03092929
2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 2-fluoro-5-nitrobenzeneboronic acid, 5-nitro-2-fluorophenylboronic acid, boronic acid, 2-fluoro-5-nitrophenyl, 2-fluoro-5-nitro-phenyl boronic acid, acmc-209pmq, ksc649e7n, 2-fluoro-5-nitro phenyl boronic acid, b-2-fluoro-5-nitrophenyl-boronic acid
(2-fluoro-5-nitrophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents