Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-nitrobenzaldehyde, 98%

$106.08 - $358.61

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-nitrobenzaldehyde
CAS 27996-87-8
Molecular Formula C7H4FNO3
Formula Weight 169.11
MDL Number MFCD00042298
Beilstein 1950329
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1923303

  SDS
alfa aesar™
L1923303
1g Each for $106.08

AAL1923306

  SDS
alfa aesar™
L1923306
5g Each for $358.61
Description & Specifications

Specifications

Formula Weight 169.11
CAS 27996-87-8
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H4FNO3
Melting Point 57° to 59°C
Chemical Name or Material 2-Fluoro-5-nitrobenzaldehyde
MDL Number MFCD00042298
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 1g
Beilstein 1950329

Applications
2-Fluoro-5-nitrobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.