Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-nitroaniline, 98+%

$56.36 - $165.74

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-nitroaniline
CAS 369-36-8
Molecular Formula C6H5FN2O2
Formula Weight 156.12
MDL Number MFCD00007652
Beilstein 2803491
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1754514

  SDS
alfa aesar™
A1754514
25g Each for $56.36

AAA1754522

  SDS
alfa aesar™
A1754522
100g Each for $165.74
Description & Specifications

Specifications

Formula Weight 156.12
Chemical Name or Material 2-Fluoro-5-nitroaniline
Assay ≥98%
Molecular Formula C6H5FN2O2
MDL Number MFCD00007652
CAS 369-36-8
Beilstein 2803491
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 91°C (195°F)
Melting Point 99° to 103°C
Quantity 25g

Applications
It is employed as pharmaceutical intermediate.

Solubility
Practically insoluble in water.

Notes
Keep away from oxidizing agents, acid chlorides and acid anhydrides. Keep the container tightly closed in a cool, dry place. Keep away from ignition sources.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.