missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-nitroaniline, 98+%

Manufacturer:  Alfa Aesar™ 4396206

 View more versions of this product

Catalog No. AA4396206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Fluoro-5-nitroaniline
5g
C6H5FN2O2
UN2811
KJVBJICWGQIMOZ-UHFFFAOYSA-N
2-fluoro-5-nitroaniline
67785
Powder
99°C to 101°C
369-36-8
MFCD00007652
benzenamine, 2-fluoro-5-nitro, 2-fluoro-5-nitrophenylamine, 2-fluoro-5-nitro-phenylamine, 1-amino-2-fluoro-5-nitrobenzene, 2-fluoro-5-nitrobenzenamine, 2-fluoro-5-nitro aniline, 3-amino-4-fluoronitrobenzene, pubchem8504, acmc-1crtl, 5-nitro-2-fluoroaniline
C1=CC(=C(C=C1[N+](=O)[O-])N)F
156.116
156.12
≥98%

Chemical Identifiers

369-36-8
156.116
KJVBJICWGQIMOZ-UHFFFAOYSA-N
67785
C1=CC(=C(C=C1[N+](=O)[O-])N)F
C6H5FN2O2
MFCD00007652
benzenamine, 2-fluoro-5-nitro, 2-fluoro-5-nitrophenylamine, 2-fluoro-5-nitro-phenylamine, 1-amino-2-fluoro-5-nitrobenzene, 2-fluoro-5-nitrobenzenamine, 2-fluoro-5-nitro aniline, 3-amino-4-fluoronitrobenzene, pubchem8504, acmc-1crtl, 5-nitro-2-fluoroaniline
2-fluoro-5-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents