missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-[N-methyl-N-(2-nitrophenyl)carbamoyl]benzeneboronic acid, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54928MD

 View more versions of this product

Catalog No. AAH54928MD



Specifications

Specifications

2-Fluoro-5-[N-methyl-N-(2-nitrophenyl)carbamoyl]benzeneboronic acid
1449131-87-6
MFCD20265378
UOLFNPGGUUSEKJ-UHFFFAOYSA-N
[2-fluoro-5-[methyl-(2-nitrophenyl)carbamoyl]phenyl]boronic acid
73996036
97%
250mg
C14H12BFN2O5
2-fluoro-5-methyl 2-nitrophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])F)(O)O
318.067
318.07

Chemical Identifiers

1449131-87-6
318.067
UOLFNPGGUUSEKJ-UHFFFAOYSA-N
73996036
B(C1=C(C=CC(=C1)C(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])F)(O)O
C14H12BFN2O5
MFCD20265378
2-fluoro-5-methyl 2-nitrophenyl carbamoyl phenylboronic acid
[2-fluoro-5-[methyl-(2-nitrophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents