missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-[N-methyl-N-(2-nitrophenyl)carbamoyl]benzeneboronic acid, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5492803

 View more versions of this product

Catalog No. AAH5492803



Specifications

Specifications

2-Fluoro-5-[N-methyl-N-(2-nitrophenyl)carbamoyl]benzeneboronic acid
1449131-87-6
MFCD20265378
UOLFNPGGUUSEKJ-UHFFFAOYSA-N
[2-fluoro-5-[methyl-(2-nitrophenyl)carbamoyl]phenyl]boronic acid
73996036
97%
1g
C14H12BFN2O5
2-fluoro-5-methyl 2-nitrophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])F)(O)O
318.067
318.07

Chemical Identifiers

1449131-87-6
318.067
UOLFNPGGUUSEKJ-UHFFFAOYSA-N
73996036
B(C1=C(C=CC(=C1)C(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])F)(O)O
C14H12BFN2O5
MFCD20265378
2-fluoro-5-methyl 2-nitrophenyl carbamoyl phenylboronic acid
[2-fluoro-5-[methyl-(2-nitrophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents