Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-methylpyridine, 99%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-methylpyridine
CAS 2369-19-9
Molecular Formula C6H6FN
Formula Weight 111.12
MDL Number MFCD00041225
UN Number UN1993
Beilstein 107085
Synonym 6-Fluoro-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1224103

  SDS
alfa aesar™
L1224103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1224106

  SDS
alfa aesar™
L1224106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 111.12
Chemical Name or Material 2-Fluoro-5-methylpyridine
Molecular Formula C6H6FN
Assay 99%
MDL Number MFCD00041225
CAS 2369-19-9
UN Number UN1993
Synonym 6-Fluoro-3-picoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 40°C (104°F)
Beilstein 107085
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 153° to 155°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.