missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-methylpyridine, 99%

Manufacturer:  Alfa Aesar™ L1224103

 View more versions of this product

Catalog No. AAL1224103



Specifications

Specifications

2-Fluoro-5-methylpyridine
153°C to 155°C
2369-19-9
MFCD00041225
107085
AOSOZARHUJMBLZ-UHFFFAOYSA-N
2-fluoro-5-methylpyridine
2774635
99%
40°C (104°F)
1g
C6H6FN
UN1993
2-fluoro-5-picoline, 6-fluoro-3-picoline, 2-fluoro-5-methypyridine, 2-fluoro-5-methyl-pyridine, fluoromethylpyridine3, 2-fluoro-5-methyl pyridine, pyridine, 2-fluoro-5-methyl, pubchem4059, acmc-1cdos, 5-methyl-2-fluoropyridine
CC1=CN=C(C=C1)F
111.119
111.12

Chemical Identifiers

2369-19-9
111.119
AOSOZARHUJMBLZ-UHFFFAOYSA-N
2774635
CC1=CN=C(C=C1)F
C6H6FN
MFCD00041225
2-fluoro-5-picoline, 6-fluoro-3-picoline, 2-fluoro-5-methypyridine, 2-fluoro-5-methyl-pyridine, fluoromethylpyridine3, 2-fluoro-5-methyl pyridine, pyridine, 2-fluoro-5-methyl, pubchem4059, acmc-1cdos, 5-methyl-2-fluoropyridine
2-fluoro-5-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents