missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(methylcarbamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5317803

 View more versions of this product

Catalog No. AAH5317803



Specifications

Specifications

2-Fluoro-5-(methylcarbamoyl)benzeneboronic acid
1g
C8H9BFNO3
2-fluoro-5-methylcarbamoyl phenyl boronic acid, n-methyl 3-borono-4-fluorobenzamide, 2-fluoro-5-methylcarbamoyl phenylboronic acid, 2-fluoro-5-methylcarbamoyl benzeneboronic acid, 2-fluoro-5-methylcarbamoyl phenyl boronic acid, boronic acid,b-2-fluoro-5-methylamino carbonyl phenyl, acmc-209qmi, n-methyl-3-borono-4-fluorobenzamide, 2-fluoro-5-methylcarbamoyl phenyl boronicacid, 2-fluoranyl-5-methylcarbamoyl phenyl boronic acid
B(C1=C(C=CC(=C1)C(=O)NC)F)(O)O
196.972
196.97
174°C to 176°C
874289-40-4
MFCD08436028
YOQRSSHEHKHAGS-UHFFFAOYSA-N
[2-fluoro-5-(methylcarbamoyl)phenyl]boronic acid
44717464
98%

Chemical Identifiers

874289-40-4
196.972
YOQRSSHEHKHAGS-UHFFFAOYSA-N
44717464
B(C1=C(C=CC(=C1)C(=O)NC)F)(O)O
C8H9BFNO3
MFCD08436028
2-fluoro-5-methylcarbamoyl phenyl boronic acid, n-methyl 3-borono-4-fluorobenzamide, 2-fluoro-5-methylcarbamoyl phenylboronic acid, 2-fluoro-5-methylcarbamoyl benzeneboronic acid, 2-fluoro-5-methylcarbamoyl phenyl boronic acid, boronic acid,b-2-fluoro-5-methylamino carbonyl phenyl, acmc-209qmi, n-methyl-3-borono-4-fluorobenzamide, 2-fluoro-5-methylcarbamoyl phenyl boronicacid, 2-fluoranyl-5-methylcarbamoyl phenyl boronic acid
[2-fluoro-5-(methylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents