Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-methylbenzoic acid, 99%

$33.55 - $95.28

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-methylbenzoic acid
CAS 321-12-0
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD00092819
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2609403

 
alfa aesar™
H2609403
1g Each for $95.28

AAH26094MD

 
alfa aesar™
H26094MD
250mg Each for $33.55
Description & Specifications

Specifications

Formula Weight 154.14
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 160° to 162°C
Chemical Name or Material 2-Fluoro-5-methylbenzoic acid
Assay 99%
Molecular Formula C8H7FO2
MDL Number MFCD00092819
CAS 321-12-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.