missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-methylbenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

CAS 1192045-84-3
Molecular Formula C13H18BFO2
Molecular Weight (g/mol) 236.093
InChI Key XPBIIDZQZBLHMB-UHFFFAOYSA-N
Synonym 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzene
PubChem CID 56973226
IUPAC Name 2-(2-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)C)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62032MD View Documents Alfa Aesar™
H62032MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6203203 View Documents Alfa Aesar™
H6203203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1192045-84-3
236.093
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzene
2-(2-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C13H18BFO2
XPBIIDZQZBLHMB-UHFFFAOYSA-N
56973226
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)C)F

Specifications

250mg
C13H18BFO2
XPBIIDZQZBLHMB-UHFFFAOYSA-N
2-(2-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
56973226
Solid
2-Fluoro-5-methylbenzeneboronic acid pinacol ester
1192045-84-3
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzene
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)C)F
236.093
236.09
96%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures