Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-methylbenzaldehyde, 97%

$113.01 - $453.54

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-methylbenzaldehyde
CAS 93249-44-6
Molecular Formula C8H7FO
Formula Weight 138.14
MDL Number MFCD02093762
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2666103

  SDS
alfa aesar™
H2666103
1g Each for $113.01

AAH2666106

  SDS
alfa aesar™
H2666106
5g Each for $453.54
Description & Specifications

Specifications

Formula Weight 138.14
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Flash Point 178°C (352°F)
Health Hazard 1 Warning
Chemical Name or Material 2-Fluoro-5-methylbenzaldehyde
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 93249-44-6
Molecular Formula C8H7FO
Boiling Point 89°C (15mmHg)
MDL Number MFCD02093762
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.