Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-methylanisole, 96%

$55.11 - $193.95

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-methylanisole
CAS 63762-78-7
Molecular Formula C8H9FO
Formula Weight 140.16
MDL Number MFCD00665791
Beilstein 1862329
Synonym 4-Fluoro-3-methoxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1818203

 
alfa aesar™
L1818203
1g Each for $55.11

AAL1818206

 
alfa aesar™
L1818206
5g Each for $193.95
Description & Specifications

Specifications

Formula Weight 140.16
Molecular Formula C8H9FO
Beilstein 1862329
Health Hazard 1 Warning
Flash Point 71°C (159°F)
Boiling Point 93° to 95°C (30mmHg)
Chemical Name or Material 2-Fluoro-5-methylanisole
MDL Number MFCD00665791
CAS 63762-78-7
Synonym 4-Fluoro-3-methoxytoluene
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 96%
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.