missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-5-methylanisole, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L1818206
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Fluoro-5-methylanisole | |
1.07 | |
71°C (159°F) | |
1.4953 | |
5 g | |
2-fluoro-5-methylanisole, 1-fluoro-2-methoxy-4-methyl-benzene, 4-fluoro-3-methoxytoluene, benzene,1-fluoro-2-methoxy-4-methyl, benzene, 1-fluoro-2-methoxy-4-methyl, pubchem8487, acmc-1b3ll, ksc619q2t, buttpark 14\01-68 | |
COC1=C(F)C=CC(C)=C1 | |
140.16 | |
140.16 |
63762-78-7 | |
93°C to 95°C (30 mmHg) | |
C8H9FO | |
MFCD00665791 | |
1862329 | |
MOFZLPPSEOEXNJ-UHFFFAOYSA-N | |
1-fluoro-2-methoxy-4-methylbenzene | |
2737372 | |
96% |
Chemical Identifiers
63762-78-7 | |
140.16 | |
MOFZLPPSEOEXNJ-UHFFFAOYSA-N | |
1-fluoro-2-methoxy-4-methylbenzene |
C8H9FO | |
MFCD00665791 | |
2737372 | |
COC1=C(F)C=CC(C)=C1 |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures