Please login to your online account to display your discounted pricing

2-Fluoro-5-methylaniline, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  276150250

 View more versions of this product

Catalog No. AC276150250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 125.15
Quantity 25g
Color Clear yellow to brown
Density 1.109g/mL
CAS Max % 100.0
Chemical Name or Material 2-Fluoro-5-methylaniline
Storage Information Product darkens on storage
Assay Percent Range 99%
Linear Formula FC6H3(CH3)nH2
MDL Number MFCD00007654
CAS 452-84-6
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
Molecular Formula C7H8FN
Synonym 2-Fluoro-5-methylbenzeneamine
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Fluoro-5-methylaniline
CAS 452-84-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8FN
Linear Formula FC6H3(CH3)nH2
Formula Weight 125.15
MDL Number MFCD00007654
Synonym 2-Fluoro-5-methylbenzeneamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.

SDS & Safety Documents