missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-methylaniline, 99%, ACROS Organics™

Chemical Identifiers

CAS 452-84-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00007654
InChI Key QZUXMXZNVAJNSE-UHFFFAOYSA-N
Synonym 3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
PubChem CID 262970
IUPAC Name 2-fluoro-5-methylaniline
SMILES CC1=CC(=C(C=C1)F)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC276150050 SDS View Documents Acros Organics
AC276150050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

452-84-6
125.146
QZUXMXZNVAJNSE-UHFFFAOYSA-N
262970
CC1=CC(=C(C=C1)F)N
C7H8FN
MFCD00007654
3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
2-fluoro-5-methylaniline

Specifications

1.109g/mL
Glass bottle
Yellow to Brown
452-84-6
100.0
C7H8FN
MFCD00007654
QZUXMXZNVAJNSE-UHFFFAOYSA-N
2-fluoro-5-methylaniline
262970
Liquid
2-Fluoro-5-methylaniline
Authentic
1.5320 to 1.5340 (20°C, 589nm)
5g
98.5
99%
FC6H3(CH3)nH2
3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
CC1=CC(=C(C=C1)F)N
125.146
125.15
≥98.5%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : Product darkens on storage