missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-methylaniline, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC276150050

 View more versions of this product

Catalog No. AC276150050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Fluoro-5-methylaniline
81°C
Glass bottle
1.109
Yellow to Brown
452-84-6
100.0
C7H8FN
MFCD00007654
Solubility in water:
CC1=CC(=C(C=C1)F)N
125.15
125.15
≥98.5%
1.1090g/mL
Authentic
1.5320 to 1.534
80°C to 86°C (11.0mmHg)
5g
98.5
99%
FC6H3(CH3)NH2
2-fluoro-5-methyl-aniline
QZUXMXZNVAJNSE-UHFFFAOYSA-N
2-fluoro-5-methylaniline
262970
Liquid

Chemical Identifiers

452-84-6
125.15
QZUXMXZNVAJNSE-UHFFFAOYSA-N
262970
CC1=CC(=C(C=C1)F)N
C7H8FN
MFCD00007654
2-fluoro-5-methyl-aniline
2-fluoro-5-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : Product darkens on storage

SDS Documents