missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-5-methyl-4-trifluoromethyl-DL-phenylalanine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 1435806-24-8 |
---|---|
Molecular Formula | C11H11F4NO2 |
Molecular Weight (g/mol) | 265.208 |
MDL Number | MFCD19687203 |
InChI Key | ZAEBGVLZTPXBJV-UHFFFAOYSA-N |
Synonym | 2-fluoro-5-methyl-4-trifluoromethyl-dl-phenylalanine, 2-amino-3-2-fluoro-5-methyl-4-trifluoromethyl phenyl propanoic acid |
PubChem CID | 66523503 |
IUPAC Name | 2-amino-3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid |
SMILES | CC1=CC(=C(C=C1C(F)(F)F)F)CC(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AAH3384503
|
Thermo Scientific Chemicals
H3384503 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1435806-24-8 | |
265.208 | |
ZAEBGVLZTPXBJV-UHFFFAOYSA-N | |
66523503 | |
CC1=CC(=C(C=C1C(F)(F)F)F)CC(C(=O)O)N |
C11H11F4NO2 | |
MFCD19687203 | |
2-fluoro-5-methyl-4-trifluoromethyl-dl-phenylalanine, 2-amino-3-2-fluoro-5-methyl-4-trifluoromethyl phenyl propanoic acid | |
2-amino-3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid |
Specifications
1435806-24-8 | |
MFCD19687203 | |
ZAEBGVLZTPXBJV-UHFFFAOYSA-N | |
2-amino-3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid | |
66523503 | |
97% | |
2-Fluoro-5-methyl-4-trifluoromethyl-DL-phenylalanine |
C11H11F4NO2 | |
2-fluoro-5-methyl-4-trifluoromethyl-dl-phenylalanine, 2-amino-3-2-fluoro-5-methyl-4-trifluoromethyl phenyl propanoic acid | |
CC1=CC(=C(C=C1C(F)(F)F)F)CC(C(=O)O)N | |
265.208 | |
265.2 | |
1g |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures