missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-methoxyphenylboronic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445450010

 View more versions of this product

Catalog No. AC445450010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Fluoro-5-methoxyphenylboronic acid
94 % min. consistent with structure (1H-NMR)
White to Yellow
1g
94% min. (HPLC)
2-fluoro-5-methoxy-phenyl boronic acid, 2-fluoro-5-methoxybenzeneboronic acid, 2-fluoro-5-methoxyphenyl boronic acid, 2-fluoro-5-methoxyphenylboronic acid, 2-fluoro-5-methoxyphenylboronicacid, 3-borono-4-fluoroanisole, acmc-1aruy, boronic acid, 2-fluoro-5-methoxyphenyl, ksc494s2n, pubchem4080
169.95
169.95
95%
Authentic
Glass Bottle
195°C to 196°C
406482-19-7
C7H8BFO3
IPTZOWYBCLEBOE-UHFFFAOYSA-N
4157211
Solid

Chemical Identifiers

406482-19-7
169.95
2-fluoro-5-methoxy-phenyl boronic acid, 2-fluoro-5-methoxybenzeneboronic acid, 2-fluoro-5-methoxyphenyl boronic acid, 2-fluoro-5-methoxyphenylboronic acid, 2-fluoro-5-methoxyphenylboronicacid, 3-borono-4-fluoroanisole, acmc-1aruy, boronic acid, 2-fluoro-5-methoxyphenyl, ksc494s2n, pubchem4080
C7H8BFO3
IPTZOWYBCLEBOE-UHFFFAOYSA-N
4157211
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents