missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-methoxyphenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 406482-19-7
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
InChI Key IPTZOWYBCLEBOE-UHFFFAOYSA-N
Synonym 2-fluoro-5-methoxy-phenyl boronic acid
PubChem CID 4157211
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445450010 SDS View Documents Thermo Fisher Scientific
AC445450010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445450050 SDS View Documents Thermo Fisher Scientific
AC445450050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

406482-19-7
169.95
2-fluoro-5-methoxy-phenyl boronic acid
C7H8BFO3
IPTZOWYBCLEBOE-UHFFFAOYSA-N
4157211

Specifications

Authentic
Glass Bottle
195°C to 196°C
406482-19-7
C7H8BFO3
IPTZOWYBCLEBOE-UHFFFAOYSA-N
4157211
Solid
2-Fluoro-5-methoxyphenylboronic acid
94 % min. consistent with structure (1H-NMR)
White to Yellow
1g
94% min. (HPLC)
2-fluoro-5-methoxy-phenyl boronic acid
169.95
169.95
95%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning