Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester
CAS 944317-66-2
Formula Weight 294.13
MDL Number MFCD12026092
Synonym 2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6278603

 
alfa aesar™
H6278603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62786MD

 
alfa aesar™
H62786MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 294.13
Chemical Name or Material 2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
MDL Number MFCD12026092
CAS 944317-66-2
Synonym 2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester
Quantity 1g