Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-iodotoluene, 99%

$81.22 - $298.34

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-iodotoluene
CAS 452-68-6
Molecular Formula C7H6FI
Formula Weight 236.03
MDL Number MFCD00013710
Beilstein 2433078
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0839603

  SDS
alfa aesar™
L0839603
1g Each for $81.22

AAL0839606

  SDS
alfa aesar™
L0839606
5g Each for $298.34
Description & Specifications

Specifications

Formula Weight 236.03
MDL Number MFCD00013710
Chemical Name or Material 2-Fluoro-5-iodotoluene
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 99%
Quantity 1g
Health Hazard 1 Warning
CAS 452-68-6
Molecular Formula C7H6FI
Boiling Point 86° to 87°C (9mmHg)
Beilstein 2433078
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.