missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-iodopyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 171197-80-1
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.99
InChI Key GYZNHUNWABYAPO-UHFFFAOYSA-N
Synonym 2-fluoro-5-iodo pyridine, 2-fluoro-5-iodo-pyridine, 2-fluoro-5-iodopyridine, 2-fluoro-5-iodopyridine 1g, 5-iodo-2-fluoropyridine, abbypharma ap-30-0927, acmc-1bao4, ksc493k0d, pubchem6324, pyridine, 2-fluoro-5-iodo
PubChem CID 2783179
IUPAC Name 2-fluoro-5-iodopyridine
SMILES C1=CC(=NC=C1I)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441840010
SDS View Documents
Thermo Fisher Scientific
AC441840010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441840050
SDS View Documents
Thermo Fisher Scientific
AC441840050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

171197-80-1
222.99
2-fluoro-5-iodo pyridine, 2-fluoro-5-iodo-pyridine, 2-fluoro-5-iodopyridine, 2-fluoro-5-iodopyridine 1g, 5-iodo-2-fluoropyridine, abbypharma ap-30-0927, acmc-1bao4, ksc493k0d, pubchem6324, pyridine, 2-fluoro-5-iodo
2-fluoro-5-iodopyridine
C5H3FIN
GYZNHUNWABYAPO-UHFFFAOYSA-N
2783179
C1=CC(=NC=C1I)F

Specifications

95°C
Glass Bottle
35°C to 36°C
171197-80-1
100.0
C5H3FIN
GYZNHUNWABYAPO-UHFFFAOYSA-N
2-fluoro-5-iodopyridine
2783179
Crystalline Solid or Low Melting Solid
2-Fluoro-5-iodopyridine
Authentic
White to Yellow
1g
96.0
96% min. (GC)
2-fluoro-5-iodo pyridine, 2-fluoro-5-iodo-pyridine, 2-fluoro-5-iodopyridine, 2-fluoro-5-iodopyridine 1g, 5-iodo-2-fluoropyridine, abbypharma ap-30-0927, acmc-1bao4, ksc493k0d, pubchem6324, pyridine, 2-fluoro-5-iodo
C1=CC(=NC=C1I)F
222.99
222.99
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

SDS