missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-5-iodopyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441840010
Specifications
2-Fluoro-5-iodopyridine | |
Authentic | |
White to Yellow | |
1g | |
96.0 | |
96% min. (GC) | |
2-fluoro-5-iodo pyridine, 2-fluoro-5-iodo-pyridine, 2-fluoro-5-iodopyridine, 2-fluoro-5-iodopyridine 1g, 5-iodo-2-fluoropyridine, abbypharma ap-30-0927, acmc-1bao4, ksc493k0d, pubchem6324, pyridine, 2-fluoro-5-iodo | |
C1=CC(=NC=C1I)F | |
222.99 | |
222.99 | |
97% |
95°C | |
Glass Bottle | |
35°C to 36°C | |
171197-80-1 | |
100.0 | |
C5H3FIN | |
GYZNHUNWABYAPO-UHFFFAOYSA-N | |
2-fluoro-5-iodopyridine | |
2783179 | |
Crystalline Solid or Low Melting Solid |
Chemical Identifiers
171197-80-1 | |
222.99 | |
2-fluoro-5-iodo pyridine, 2-fluoro-5-iodo-pyridine, 2-fluoro-5-iodopyridine, 2-fluoro-5-iodopyridine 1g, 5-iodo-2-fluoropyridine, abbypharma ap-30-0927, acmc-1bao4, ksc493k0d, pubchem6324, pyridine, 2-fluoro-5-iodo | |
2-fluoro-5-iodopyridine |
C5H3FIN | |
GYZNHUNWABYAPO-UHFFFAOYSA-N | |
2783179 | |
C1=CC(=NC=C1I)F |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning