Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-iodopyridine, 97%

$47.72 - $198.57

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-iodopyridine
CAS 171197-80-1
Molecular Formula C5H3FIN
Formula Weight 222.99
MDL Number MFCD03095301
Beilstein 7756788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2004103

 
alfa aesar™
L2004103
1g Each for $47.72

AAL2004106

 
alfa aesar™
L2004106
5g Each for $198.57
Description & Specifications

Specifications

Formula Weight 222.99
Chemical Name or Material 2-Fluoro-5-iodopyridine
Health Hazard 1 Warning
Assay 97%
MDL Number MFCD03095301
CAS 171197-80-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C5H3FIN
Beilstein 7756788
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 31° to 33°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.