missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-hydroxypyridine, 95%

$464.86 - $464.86

Chemical Identifiers

CAS 55758-32-2
Molecular Formula C5H4FNO
Formula Weight 113.09
MDL Number MFCD01075040
Synonym 6-Fluoro-3-pyridinol
Chemical Name or Material 2-Fluoro-5-hydroxypyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2774103 SDS Alfa Aesar™
H2774103
1g Each for $464.86
AAH2774106 SDS Alfa Aesar™
H2774106
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

55758-32-2
113.09
6-Fluoro-3-pyridinol
C5H4FNO
MFCD01075040
2-Fluoro-5-hydroxypyridine

Specifications

150° to 153°C
55758-32-2
113.09
6-Fluoro-3-pyridinol
95%
1g
C5H4FNO
MFCD01075040
2-Fluoro-5-hydroxypyridine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning