Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(hydroxymethyl)benzeneboronic acid, 98%

$50.65 - $247.82

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-(hydroxymethyl)benzeneboronic acid
CAS 1072952-25-0
Molecular Formula C7H8BFO3
Formula Weight 169.95
MDL Number MFCD06656276
Synonym 3-Borono-4-fluorobenzyl alcohol; 2-Fluoro-5-(hydroxymethyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2925103

 
alfa aesar™
H2925103
1g Each for $50.65

AAH2925109

 
alfa aesar™
H2925109
10g Each for $247.82
Description & Specifications

Specifications

Formula Weight 169.95
Chemical Name or Material 2-Fluoro-5-(hydroxymethyl)benzeneboronic acid
Assay 98%
Molecular Formula C7H8BFO3
MDL Number MFCD06656276
CAS 1072952-25-0
Synonym 3-Borono-4-fluorobenzyl alcohol; 2-Fluoro-5-(hydroxymethyl)phenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 64° to 76°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.