Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-hydroxybenzoic acid, 98%

$116.77 - $292.31

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-hydroxybenzoic acid
CAS 51446-30-1
Molecular Formula C7H5FO3
Formula Weight 156.11
MDL Number MFCD01862084
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2615003

  SDS
alfa aesar™
H2615003
1g Each for $292.31

AAH26150MD

  SDS
alfa aesar™
H26150MD
250mg Each for $116.77
Description & Specifications

Specifications

Formula Weight 156.11
Health Hazard 1 Warning
Melting Point 204° to 206°C
CAS 51446-30-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 2-Fluoro-5-hydroxybenzoic acid
MDL Number MFCD01862084
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H5FO3
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.