missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445030010

 View more versions of this product

Catalog No. AC445030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Fluoro-5-formylphenylboronic acid pinacol ester
105°C to 113°C (0.1mmHg)
1g
94% min. (GC)
2-fluoro-5-formylbenzeneboronic acid pinacol ester, 2-fluoro-5-formylphenyl boronic acid pinacol ester, 2-fluoro-5-formylphenylboronic acid pinacol ester, 2-fluoro-5-formylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzaldehyde, benzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
250.08
250.08
Glass Bottle
32°C to 37°C
1112208-82-8
C13H16BFO3
WHYMLQVABFAGEK-UHFFFAOYSA-N
46738018
95%

Chemical Identifiers

1112208-82-8
250.08
2-fluoro-5-formylbenzeneboronic acid pinacol ester, 2-fluoro-5-formylphenyl boronic acid pinacol ester, 2-fluoro-5-formylphenylboronic acid pinacol ester, 2-fluoro-5-formylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzaldehyde, benzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
C13H16BFO3
WHYMLQVABFAGEK-UHFFFAOYSA-N
46738018
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents