Please login to your online account to display your discounted pricing

2-Fluoro-5-formylbenzonitrile, 99%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC397940050

 View more versions of this product

Catalog No. AC397940050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 149.12
Quantity 5g
Molecular Formula C8H4FNO
Linear Formula FC6H3(CN)CHO
CAS 218301-22-5
Chemical Name or Material 2-Fluoro-5-formylbenzonitrile
Assay Percent Range 99%
MDL Number MFCD01863558

Specifications

Chemical Name or Material 2-Fluoro-5-formylbenzonitrile
Assay Percent Range 99%
Molecular Formula C8H4FNO
Linear Formula FC6H3(CN)CHO
Formula Weight 149.12
MDL Number MFCD01863558