Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-formylbenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-formylbenzeneboronic acid pinacol ester
CAS 1112208-82-8
Formula Weight 250.07
MDL Number MFCD11867877
Synonym 2-Fluoro-5-formylphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6268903

 
alfa aesar™
H6268903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6268906

 
alfa aesar™
H6268906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62689MD

 
alfa aesar™
H62689MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.07
Melting Point 35° to 40°C
Chemical Name or Material 2-Fluoro-5-formylbenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
MDL Number MFCD11867877
CAS 1112208-82-8
Synonym 2-Fluoro-5-formylphenylboronic acid pinacol ester
Quantity 1g