Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-formylbenzeneboronic acid, 97%

$175.47 - $527.97

Chemical Identifiers

Chemical Name or Material 2-Fluoro-5-formylbenzeneboronic acid
CAS 352534-79-3
Molecular Formula C7H6BFO3
Formula Weight 167.93
MDL Number MFCD03095100
Synonym 3-Borono-4-fluorobenzaldehyde; 2-Fluoro-5-formylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2885703

 
alfa aesar™
H2885703
1g Each for $175.47

AAH2885706

 
alfa aesar™
H2885706
5g Each for $527.97
Description & Specifications

Specifications

Formula Weight 167.93
Chemical Name or Material 2-Fluoro-5-formylbenzeneboronic acid
Assay 97%
Molecular Formula C7H6BFO3
MDL Number MFCD03095100
CAS 352534-79-3
Synonym 3-Borono-4-fluorobenzaldehyde; 2-Fluoro-5-formylphenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 168° to 171°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.