missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(4-morpholinylcarbonyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5421303

 View more versions of this product

Catalog No. AAH5421303



Specifications

Specifications

2-Fluoro-5-(4-morpholinylcarbonyl)benzeneboronic acid
1072951-41-7
MFCD11504847
PGPWWCUYTSKDEA-UHFFFAOYSA-N
[2-fluoro-5-(morpholine-4-carbonyl)phenyl]boronic acid
46738991
97%
1g
C11H13BFNO4
2-fluoro-5-4-morpholinylcarbonyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)N2CCOCC2)F)(O)O
253.036
253.04

Chemical Identifiers

1072951-41-7
253.036
PGPWWCUYTSKDEA-UHFFFAOYSA-N
46738991
B(C1=C(C=CC(=C1)C(=O)N2CCOCC2)F)(O)O
C11H13BFNO4
MFCD11504847
2-fluoro-5-4-morpholinylcarbonyl benzeneboronic acid
[2-fluoro-5-(morpholine-4-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents