Alfa Aesar™ 2-Fluoro-5-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Manufacturer:  Alfa Aesar™ H54985MD
 View more versions of this product
Catalog No. AAH54985MD
Specifications
Specifications
2-Fluoro-5-(4-fluorobenzylcarbamoyl)benzeneboronic acid
2096333-62-7
C14H12BF2NO3
2-Fluoro-5-(4-fluorobenzylcarbamoyl)phenylboronic acid
291.06
291.06
250mg
97%
MFCD20040282
MIWOFLMSLFRZDO-UHFFFAOYSA-N
73996067
Chemical Identifiers
2096333-62-7
291.06
MIWOFLMSLFRZDO-UHFFFAOYSA-N
73996067
C14H12BF2NO3
MFCD20040282
2-Fluoro-5-(4-fluorobenzylcarbamoyl)phenylboronic acid
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
SDS Documents