missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid, 97%

$180.00 - $573.77

Chemical Identifiers

CAS 1449132-32-4
Molecular Formula C12H10BFN2O3
Molecular Weight (g/mol) 260.031
MDL Number MFCD20265175
InChI Key LWVDXBKALYJJIU-UHFFFAOYSA-N
Synonym 2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid
PubChem CID 73995628
IUPAC Name [2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54093MD View Documents Alfa Aesar™
H54093MD
250mg Each for $180.00
 
AAH5409303 View Documents Alfa Aesar™
H5409303
1g Each for $573.77
 
Specifications

Chemical Identifiers

1449132-32-4
260.031
LWVDXBKALYJJIU-UHFFFAOYSA-N
73995628
B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
C12H10BFN2O3
MFCD20265175
2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid
[2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid

Specifications

250mg
C12H10BFN2O3
2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
260.031
260.03
2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid
1449132-32-4
MFCD20265175
LWVDXBKALYJJIU-UHFFFAOYSA-N
[2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid
73995628
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures