missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5409303

 View more versions of this product

Catalog No. AAH5409303



Specifications

Specifications

2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid
1449132-32-4
MFCD20265175
LWVDXBKALYJJIU-UHFFFAOYSA-N
[2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid
73995628
97%
1g
C12H10BFN2O3
2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
260.031
260.03

Chemical Identifiers

1449132-32-4
260.031
LWVDXBKALYJJIU-UHFFFAOYSA-N
73995628
B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
C12H10BFN2O3
MFCD20265175
2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid
[2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents