missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(3-methylphenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54903MD

 View more versions of this product

Catalog No. AAH54903MD



Specifications

Specifications

2-Fluoro-5-(3-methylphenylcarbamoyl)benzeneboronic acid
1449135-55-0
MFCD20265368
DULJUFCUHPACGN-UHFFFAOYSA-N
[2-fluoro-5-[(3-methylphenyl)carbamoyl]phenyl]boronic acid
73996021
97%
250mg
C14H13BFNO3
2-fluoro-5-3-methylphenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC(=C2)C)F)(O)O
273.07
273.07

Chemical Identifiers

1449135-55-0
273.07
DULJUFCUHPACGN-UHFFFAOYSA-N
73996021
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC(=C2)C)F)(O)O
C14H13BFNO3
MFCD20265368
2-fluoro-5-3-methylphenyl carbamoyl phenylboronic acid
[2-fluoro-5-[(3-methylphenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents