missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(3-fluorophenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5489703

 View more versions of this product

Catalog No. AAH5489703



Specifications

Specifications

2-Fluoro-5-(3-fluorophenylcarbamoyl)benzeneboronic acid
2096336-75-1
MFCD20040266
XUFYLZBKRBENSW-UHFFFAOYSA-N
73996018
97%
1g
C13H10BF2NO3
2-fluoro-5-3-fluorophenylcarbamoyl phenylboronic acid, 2-fluoro-5-3-fluorophenylcarbamoyl benzeneboronic acid
277.03
277.03

Chemical Identifiers

2096336-75-1
277.03
XUFYLZBKRBENSW-UHFFFAOYSA-N
73996018
C13H10BF2NO3
MFCD20040266
2-fluoro-5-3-fluorophenylcarbamoyl phenylboronic acid, 2-fluoro-5-3-fluorophenylcarbamoyl benzeneboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents