missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5423003

 View more versions of this product

Catalog No. AAH5423003



Specifications

Specifications

2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)benzeneboronic acid
1449132-35-7
MFCD20040159
LZBLIQJRKZLMQM-UHFFFAOYSA-N
[2-fluoro-5-[(3-fluoro-4-methylphenyl)methylcarbamoyl]phenyl]boronic acid
73995702
97%
1g
C15H14BF2NO3
2-fluoro-5-3-fluoro-4-methylbenzylcarbamoyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)NCC2=CC(=C(C=C2)C)F)F)(O)O
305.088
305.09

Chemical Identifiers

1449132-35-7
305.088
LZBLIQJRKZLMQM-UHFFFAOYSA-N
73995702
B(C1=C(C=CC(=C1)C(=O)NCC2=CC(=C(C=C2)C)F)F)(O)O
C15H14BF2NO3
MFCD20040159
2-fluoro-5-3-fluoro-4-methylbenzylcarbamoyl benzeneboronic acid
[2-fluoro-5-[(3-fluoro-4-methylphenyl)methylcarbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents