missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(2-methyl-1-piperidinylcarbonyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5438203

 View more versions of this product

Catalog No. AAH5438203



Specifications

Specifications

2-Fluoro-5-(2-methyl-1-piperidinylcarbonyl)benzeneboronic acid
1449144-45-9
MFCD20265256
WUPGAFQQYJVDKA-UHFFFAOYSA-N
[2-fluoro-5-(2-methylpiperidine-1-carbonyl)phenyl]boronic acid
73995783
97%
1g
C13H17BFNO3
2-fluoro-5-2-methyl-1-piperidinylcarbonyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)N2CCCCC2C)F)(O)O
265.091
265.09

Chemical Identifiers

1449144-45-9
265.091
WUPGAFQQYJVDKA-UHFFFAOYSA-N
73995783
B(C1=C(C=CC(=C1)C(=O)N2CCCCC2C)F)(O)O
C13H17BFNO3
MFCD20265256
2-fluoro-5-2-methyl-1-piperidinylcarbonyl benzeneboronic acid
[2-fluoro-5-(2-methylpiperidine-1-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents