missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(2-methoxy-4-nitrophenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54224MD

 View more versions of this product

Catalog No. AAH54224MD



Specifications

Specifications

2-Fluoro-5-(2-methoxy-4-nitrophenylcarbamoyl)benzeneboronic acid
1449144-38-0
MFCD20265209
BYDYGPUXHRRBSV-UHFFFAOYSA-N
[2-fluoro-5-[(2-methoxy-4-nitrophenyl)carbamoyl]phenyl]boronic acid
73995700
97%
250mg
C14H12BFN2O6
2-fluoro-5-2-methoxy-4-nitrophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=C(C=C2)[N+](=O)[O-])OC)F)(O)O
334.066
334.07

Chemical Identifiers

1449144-38-0
334.066
BYDYGPUXHRRBSV-UHFFFAOYSA-N
73995700
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=C(C=C2)[N+](=O)[O-])OC)F)(O)O
C14H12BFN2O6
MFCD20265209
2-fluoro-5-2-methoxy-4-nitrophenyl carbamoyl phenylboronic acid
[2-fluoro-5-[(2-methoxy-4-nitrophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents