missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54510MD

 View more versions of this product

Catalog No. AAH54510MD



Specifications

Specifications

2-Fluoro-5-(2-fluorobenzylcarbamoyl)benzeneboronic acid
1449144-70-0
MFCD20040208
KQSVTPWXLNYZIX-UHFFFAOYSA-N
[2-fluoro-5-[(2-fluorophenyl)methylcarbamoyl]phenyl]boronic acid
73995846
97%
250mg
C14H12BF2NO3
2-fluoro-5-2-fluorobenzylcarbamoyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)NCC2=CC=CC=C2F)F)(O)O
291.061
291.06

Chemical Identifiers

1449144-70-0
291.061
KQSVTPWXLNYZIX-UHFFFAOYSA-N
73995846
B(C1=C(C=CC(=C1)C(=O)NCC2=CC=CC=C2F)F)(O)O
C14H12BF2NO3
MFCD20040208
2-fluoro-5-2-fluorobenzylcarbamoyl benzeneboronic acid
[2-fluoro-5-[(2-fluorophenyl)methylcarbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents