missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-5-(2-fluoro-5-nitrophenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54774MD

 View more versions of this product

Catalog No. AAH54774MD



Specifications

Specifications

2-Fluoro-5-(2-fluoro-5-nitrophenylcarbamoyl)benzeneboronic acid
1449133-65-6
MFCD20091227
MTQQOSSMDAQKRY-UHFFFAOYSA-N
[2-fluoro-5-[(2-fluoro-5-nitrophenyl)carbamoyl]phenyl]boronic acid
73995958
97%
250mg
C13H9BF2N2O5
2-fluoro-5-2-fluoro-5-nitrophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)[N+](=O)[O-])F)F)(O)O
322.031
322.03

Chemical Identifiers

1449133-65-6
322.031
MTQQOSSMDAQKRY-UHFFFAOYSA-N
73995958
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)[N+](=O)[O-])F)F)(O)O
C13H9BF2N2O5
MFCD20091227
2-fluoro-5-2-fluoro-5-nitrophenyl carbamoyl phenylboronic acid
[2-fluoro-5-[(2-fluoro-5-nitrophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents