Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Fluoro-4'-(trifluoromethyl)propiophenone, 97%

$185.84 - $185.84

Chemical Identifiers

Chemical Name or Material 2'-Fluoro-4'-(trifluoromethyl)propiophenone
CAS 208173-16-4
Molecular Formula C10H8F4O
Formula Weight 220.17g/mol
MDL Number MFCD00061259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2455206

 
alfa aesar™
B24552-06
5g Each for $185.84
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 2'-Fluoro-4'-(trifluoromethyl)propiophenone
CAS 208173-16-4
Assay 97%
Molecular Formula C10H8F4O
Formula Weight 220.17g/mol
MDL Number MFCD00061259
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.