missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)benzylamine, 97%

Manufacturer:  Alfa Aesar™ L1146203

 View more versions of this product

Catalog No. AAL1146203



Specifications

Specifications

2-Fluoro-4-(trifluoromethyl)benzylamine
239087-05-9
MFCD00070807
2-fluoro-4-trifluoromethyl benzylamine, 2-fluoro-4-trifluoromethyl phenyl methanamine, 2-fluoro-4-trifluoromethylbenzylamine, 2-fluoro-4-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl phenyl methylamine, 1-2-fluoro-4-trifluoromethyl phenyl methanamine, acmc-209uw2, ksc494q1h, 2-fluoro-4-trifluoromethyl benzylamine
C1=CC(=C(C=C1C(F)(F)F)F)CN
193.145
193.15
1g
C8H7F4N
UN2735
MQTBAGAVFDZXKF-UHFFFAOYSA-N
[2-fluoro-4-(trifluoromethyl)phenyl]methanamine
2737566
97%

Chemical Identifiers

239087-05-9
193.145
MQTBAGAVFDZXKF-UHFFFAOYSA-N
2737566
C1=CC(=C(C=C1C(F)(F)F)F)CN
C8H7F4N
MFCD00070807
2-fluoro-4-trifluoromethyl benzylamine, 2-fluoro-4-trifluoromethyl phenyl methanamine, 2-fluoro-4-trifluoromethylbenzylamine, 2-fluoro-4-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl phenyl methylamine, 1-2-fluoro-4-trifluoromethyl phenyl methanamine, acmc-209uw2, ksc494q1h, 2-fluoro-4-trifluoromethyl benzylamine
[2-fluoro-4-(trifluoromethyl)phenyl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Documents