Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)benzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzyl bromide
CAS 239087-07-1
Molecular Formula C8H5BrF4
Formula Weight 257.03
MDL Number MFCD00061173
UN Number UN1759
Synonym alpha-Bromo-2-fluoro-4-(trifluoromethyl)toluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2317203

  SDS
alfa aesar™
B2317203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2317206

  SDS
alfa aesar™
B2317206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.03
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzyl bromide
Assay 97%
Molecular Formula C8H5BrF4
MDL Number MFCD00061173
CAS 239087-07-1
UN Number UN1759
Synonym alpha-Bromo-2-fluoro-4-(trifluoromethyl)toluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Melting Point 28° to 30°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.