Please login to your online account to display your discounted pricing

2-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397820010

 View more versions of this product

Catalog No. AC397820010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 226.56
Quantity 1g
Color Clear colorless
Density 1.5g/mL
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C8H3ClF4O
Linear Formula FC6H3(CF3)COCl
CAS 126917-10-0
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
Assay Percent Range 97%
MDL Number MFCD00061155
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)

Specifications

Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
CAS 126917-10-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H3ClF4O
Linear Formula FC6H3(CF3)COCl
Formula Weight 226.56
MDL Number MFCD00061155
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents