Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)benzophenone, 97%

$44.15 - $111.19

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzophenone
CAS 207974-08-1
Molecular Formula C14H8F4O
Formula Weight 268.21
MDL Number MFCD00061253
Beilstein 8913074
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1146003

  SDS
alfa aesar™
L1146003
1g Each for $111.19

AAL11460MD

  SDS
alfa aesar™
L11460MD
250mg Each for $44.15
Description & Specifications

Specifications

Formula Weight 268.21
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzophenone
Assay 97%
Molecular Formula C14H8F4O
MDL Number MFCD00061253
CAS 207974-08-1
Beilstein 8913074
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Melting Point 38° to 40°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.